SP Bel-Art Mini Magnifier, Bel-Art Products, a part of SP
Supplier: Bel-Art Products, a Part of SP
F37865-0000
54848-306EA 416.26 USD
54848-306
SP Bel-Art Mini Magnifier, Bel-Art Products, a part of SP
Magnifiers
Freestanding magnifier permits enhanced viewing of slides, Petri dishers, multiwell plates, gels, or text.
  • Freestanding design
  • Can be placed over a variety of objects
  • Simple to use
Precision-molded lens offers 1.75 magnification. The unique stand allows the magnifier to be placed over a variety of objects.
Stand height: 17.2 cm (63/4")
Unit weight: 368.5 g (13 oz.)