SP Bel-Art Colony Counter Replacement Tips, Bel-Art Products, a part of SP
Supplier: Bel-Art Products, a Part of SP
H37862-0100
54848-298EA 41.13 USD
54848-298
SP Bel-Art Colony Counter Replacement Tips, Bel-Art Products, a part of SP
Pens Lab Markers
Replacement tips fit discontinued Scienceware® pen-style Colony Counter model F37862-0000.
  • Black markers
  • Oil-resistant
  • Water-resistant
Long-life black felt-tip markers work on plastic or glass, and are oil-and water-resistant.
Ink is removable with ethyl alcohol.