SP Bel-Art Minipet™ Pipettor, Bel-Art Products, a part of SP
Supplier: Bel-Art Products, a Part of SP
F37900-1100
54848-194EA 607.69 USD
54848-194 54848-198 54848-202
SP Bel-Art Minipet™ Pipettor, Bel-Art Products, a part of SP
Pipettes Repetitive Pipettors
Pipettors feature a three-way check valve for continuous repetitive pipetting.
  • High chemical resistance
  • Borosilicate glass and Teflon® resin components
  • Aqueous pipettors are autoclavable
Just place the filling line in a reservoir, push the Teflon® resin plunger to dispense, and release to refill.
All-Teflon® resin pipettors offer a high level of chemical resistance. They may be used with virtually any solvent or acid (except HF). Fluids come in contact with only borosilicate glass and Teflon® resin. Supplied with an all-Teflon® resin valve with glass check, a Teflon® resin sinker, and a 45.7 cm (18") Teflon® resin filling line with an Int.Ø of 3.2 mm (¹/₈").
Aqueous pipettors are autoclavable and can be used with a Luer-lock manifold for multiwell plate work. They are supplied with a chrome-plated check valve, Luer-lock fitting, Buna-N seals, stainless steel spring, and Luer lock cannula, 45.7 cm long (18"), 3.2 mm (¹/₈") Int.Ø silicon filling tube and Teflon® sinker.
A molded polypropylene handle offers comfort for extended use. Turning the knob on top sets the self-locking capacity to assure repeatable dispensing. Extensions can be added to the filling line for greater reach.