SP Bel-Art Glove Box Port Covers, 8" ext.Ø., Bel-Art Products, a part of SP
Supplier: Bel-Art Products, a Part of SP
H50025-0008
54848-128EA 317.87 USD
54848-128
SP Bel-Art Glove Box Port Covers, 8" ext.Ø., Bel-Art Products, a part of SP
Covers
Use these glove box port covers to seal off any glove ports to which gloves are not attached.
  • Single-piece design
  • Neoprene gasket provides a snug fit
  • Easy to apply port covers
Covers are one-piece construction with a neoprene gasket for a snug fit and fit glove ports with a 20.3 cm (8") O.D. Simply press the covers into the port opening until the flange on the cover meets the glove port.