Magid® Heater Beater® 97GT Cotton Canvas Hot Mill Gloves, Magid
Supplier: Magid Glove
97BT
54846-009CS 996.03 USD
54846-009
Magid® Heater Beater® 97GT Cotton Canvas Hot Mill Gloves, Magid
Gloves
The magid heater beater series is choice for heavy-duty heat gloves and standard for metal workers.
  • Heavy-duty 24 oz. weight
  • Thick triple-layer design
  • Nap out palm for improved grip, heat protection
Hot Mill gloves keep out the heat and provide superior comfort and protection. This sturdy glove has a unique triple layer design that features heat-resistant burlap between outer and inner layers of cotton for maximum heat protection and comfort.
They also feature a soft nap out palm with and a 2" band top cuff or 5" gauntlet cuff for protection past the wrist and easy on/off.