Water Heater Recirculator, Zahn Electronics
Supplier: RPR
REC-H1000
54811-021EA 4243.49 USD
54811-021
Water Heater Recirculator, Zahn Electronics
Circulating Thermostats Closed Circulator Systems Heating Circulators
Portable, closed system designed for maintaining a water bath temperature for high temperature applications.
  • Solid state controller has a holding accuracy of ±0.25°F
  • It comes equipped with a heavy duty magnetic drive pump designed to deliver continuous service
  • The pump capacity is rated to pump 480 GPH, at a 3 foot rise
  • The units are complimented by an all stainless steel cover and chasis
Ordering information: Supplied with a power cord and red indicator light.