Test Cylinder Outfit, Carver
Supplier: Carver
2090-0
53889-009EA 1824.62 USD
53889-009
Test Cylinder Outfit, Carver
Presses
For pressing out samples of vegetable oil from oil-bearing materials; molding, briquetting, and forming materials under pressure; and studying effect of various unit pressures
I.D. Outfit: 2.86cm (11/8").
Ordering information: Complete with stainless steel plunger and base plug, nickel-plated cylinder, and ejector.