Electric Hot Plate Set, Carver
Supplier: Carver
2101-1-115V
53881-007PK 4827.42 USD
53881-007
Electric Hot Plate Set, Carver
Presses
For use with standard laboratory presses (see 53880-048 series)
This hot plate set incorporates a top and bottom aluminum hot plate. These 15.2cm (6") plates have individual temperature controllers and thermometers. They provide heat from 65.6° to 260°C (150° to 500°F).