Pans, Stainless Steel, Carver
Supplier: Carver
237005
53880-010EA 254.46 USD
53880-010
Pans, Stainless Steel, Carver
Trays/Dishes/Pots Laboratory Trays/Dishes
Stainless steel fluid pan for use with all Test Cylinder Outfits.
When presses are used for fluid extraction, filter pads sandwich the sample. Stainless steel pans capture the released fluid.