SP Bel-Art Policeman Stirrer, Polypropylene, Bel-Art Products, a part of SP
Supplier: Bel-Art Products, a Part of SP
F37739-0001
53800-005CS 336.4 USD
53800-005
SP Bel-Art Policeman Stirrer, Polypropylene, Bel-Art Products, a part of SP
Spatulas
This policeman stirrer is made of a 4.7 mm (³/₁₆") Ø polypropylene rod, with one end flattened and cut diagonally.
  • Ergonomic design
  • Polypropylene rod construction
  • Suitable for scraping, transferring materials, and stirring
  • Autoclavable
Useful for scraping and transferring materials, it is steam autoclavable at 121 °C (250 °F).