SP Bel-Art Pipette Tray Rack, Bel-Art Products, a part of SP
Supplier: Bel-Art Products, a Part of SP
F18940-0000
53574-100CS 1464.53 USD
53574-100
SP Bel-Art Pipette Tray Rack, Bel-Art Products, a part of SP
Pipette Stands
This styrene tray is useful in lab drawers or on a benchtop for soaking, draining, or storing pipettes.
  • Styrene construction
  • Scalloped rims hold different pipette sizes
  • Compact design
Scalloped rims hold 16 small or 7 large pipettes. Center support holds 12 short pipettes.