Stainless Steel Pipette Boxes, Boekel Industries
Supplier: Boekel
1369S0
53570-925EA 54.62 USD
53570-925
Stainless Steel Pipette Boxes, Boekel Industries
Boxes Pipette Sterilization Containers
Used to hold and protect pipettes during sterilization
Constructed of stainless steel for high corrosion resistance. Eliminates the possibility of container rolling off the benchtop. Intended for holding Pasteur pipettes 14.6 cm to 22.9 cm long.