ART® 200G Self-Sealing Barrier Pipette Tips, Molecular BioProducts
Supplier: Thermo Scientific
2069G
53509-502CS 1184.42 USD
53509-502
ART® 200G Self-Sealing Barrier Pipette Tips, Molecular BioProducts
Pipette Tips
Designed with a patented, self-sealing barrier, ART tips are the only tips that completely block the passage of aqueous based liquids and aerosols from the sample to the pipettor.
  • All tips are pre-sterilized
ART tips are designed to draw and dispense the full volume of the tip and the fine point design is an additional feature to ensure that samples can be drawn and dispensed with accuracy.
Electron beam sterilization is used because it ensures total disruption of any molecules amplifiable by PCR.
Tips are compatible with Eppendorf®, Oxford BenchMate®, Finnpipette®, Pipetman®, and Rainin® series pipettors.
The unique seal protects the pipetting device from any possible contamination that could carry over and compromise future samples. ART tips are ideal for critical uses such as PCR, sequencing, loading gels, handling infectious samples and pipetting radioactive isotopes. The barrier formed by an ART filter is so secure it even prevents 7.2 million counts of 32P from breaking through. The purity of ART tips is achieved by constant control against contamination from any source.
Certifications: Certified RNase-, DNase-, DNA-, and pyrogen-free.
Packaging: Tips are packaged 96/tray, 10 trays per pack.