Monoject™ LIFE SHIELD™ Blunt Cannula, Covidien
Supplier: Cardinal Health
8881202017
53503-936BX 124.87 USD
53503-936
Monoject™ LIFE SHIELD™ Blunt Cannula, Covidien
Clinical Diagnostic Supplies Clinical Sample Collection/Preparation Blood Sample Collection/Preparation
This sterile, stainless steel cannula with a polypropylene luerlock hub may be used to administer fluids intravenously or draw blood through an inserted catheter.