Socorex™ Calibrex™ 520 Bottle-Top Dispensers, DWK Life Sciences
Supplier: DWK Life Sciences (KIMBLE)
844000 844002 844004
53501-140EA 1042.65 USD
53501-140 53501-142 53501-144
Socorex™ Calibrex™ 520 Bottle-Top Dispensers, DWK Life Sciences
Bottle Top Dispensers
These chemically inert bottle-top dispensers feature a series of permanently calibrated stops that provide quick and accurate volume selection
Calibration is easily adjusted in the laboratory for temperature or viscosity changes. The volume display is adjustable to four positions for optimal user viewing and ease of setting. The airflow orifice can be modified to accept a membrane filter that eliminates aerosol contamination. The syringe barrel is encased in a protective sleeve, preventing breakage and eliminating spills. The dispenser is autoclavable when fully assembled and easilyMore Product Information