Replacement Valve Assembly Kits for Self-Refilling Syringes, DWK Life Sciences
Supplier: DWK Life Sciences (KIMBLE)
851431
53500-895EA 527.96 USD
53500-895 53500-896
Replacement Valve Assembly Kits for Self-Refilling Syringes, DWK Life Sciences
Valves Injection Valves
Designed for use with Wheaton self-refilling syringes.
Delivery information: Each kit includes 2 fluoropolymer resin washers, 2 corrosion-resistant springs, 2 stainless steel balls, and Viton washer for barrel (except for 20 ml syringe).