Micro-Splitter Valve, Upchurch Scientific®
Supplier: Upchurch Scientific
P-450
53500-628EA 130.89 USD
53500-628 53500-630 53521-047 53521-049
Micro-Splitter Valve, Upchurch Scientific®
Valves Micro Valves
Designed to accurately split and control a low-flow stream of a single incoming supply.
  • For interfacing LC-MS systems
  • Pressure rated to 800 psi (55 bar)
  • Adjustable split stream flow ratesFor interfacing LC-MS systems
  • Pressure rated to 800 psi (55 bar)
  • Adjustable split stream flow ratesFor interfacing LC-MS systems
  • Pressure rated to 800 psi (55 bar)
  • Adjustable split stream flow rates