Disposable Pasteur Pipettes, Borosilicate Glass, Wheaton
Supplier: DWK Life Sciences (KIMBLE)
357331
53499-630CS 148.48 USD
53499-630
Disposable Pasteur Pipettes, Borosilicate Glass, Wheaton
Pipettes Pasteur/Transfer Pipettes
For use in inoculating culture media and as a general sampling pipet.
Also used in applying samples in chromatography. Straight tip can be bent over a flame. Constriction near end to facilitate plugging. Pipettes have an Ext.Ø of 7 ±0.5 mm at the top.
Ordering information: Case of 1000 Pasteur Pipettes. Each case includes 4 inner cartons of 250 pipettes.