Medium Pressure 4-Port Switching Valves, Upchurch Scientific®, IDEX Health & Science
Supplier: Upchurch Scientific
V-101D
53500-812EA 208.93 USD
53500-812 53498-576
Medium Pressure 4-Port Switching Valves, Upchurch Scientific®, IDEX Health & Science
Valves Switching Valves
This 4 Way PEEK Bulkhead Flow Valve is ideal if your application involves low frequency of use and demands operator control or involves injection of smaller sample volumes.
  • Biocompatible and chemically inert
  • Pressure rated to 500 psi (34 bar)