SP Bel-Art Bel-Bulb™ Pipettor, Bel-Art Products, a part of SP
Supplier: Bel-Art Products, a Part of SP
F37887-0000
53497-304CS 215.19 USD
53497-304
SP Bel-Art Bel-Bulb™ Pipettor, Bel-Art Products, a part of SP
Pipettes Pipette Bulbs
This simple suction device with a soft tapered chuck has a flexible vinyl bulb that is 5Ø×7 L cm (2×2¾").
  • Simple
  • Soft, tapered chuck
  • Flexible vinyl bulb
  • Easy to use
After filling the pipet, remove the bulb and quickly cover the top of the pipette with a gloved finger to hold fluid or dispense it as needed. Fits pipettes from 6 to 8 mm.