VWR® FLIP™ Pipette Filler
Supplier: VWR International
53497-055
53497-055CS 7049.57 USD
53497-055
VWR® FLIP™ Pipette Filler
Pipettes Pipette Bulbs
Pipette filler for all pipettes from 0.05 to 100 ml.
  • Natural rubber
  • Easy to use
Simply squeeze the ball and it is ready to use
One-piece, natural red rubber pipette filler is equipped with an automatic valve that is removable for easy cleaning. The automatic blowout valve eliminates the need for two hands to evacuate the bulb.
Press the 'up' arrow to intake liquid; press the 'down' arrow on the arm to empty. Press below the 'up' arrow to release the pipet.