KIMAX® Reusable Bacteriological Pipettes with Safe-Guard® Tempered Tip, DWK Life Sciences
Supplier: DWK Life Sciences (KIMBLE)
37081-110
53381-060CS 284.12 USD
53381-060
KIMAX® Reusable Bacteriological Pipettes with Safe-Guard® Tempered Tip, DWK Life Sciences
Pipettes Bacteriological Pipettes
KIMAX® pipette with a subdivision scale of permanent brown stain fused into the surface of uniform bore tubing without etching.
  • Tooled for a cotton plug at the top end
  • Correct volume is delivered without the need to blow out
  • Designed from ASTM Specification E1043, Type V requirements
Pipettes are manufactured from 51 expansion borosilicate glass.
Certifications: Material conforms to USP Type I and ASTM E438, Type I, Class B requirements.