Falcon® Disposable Serological Pipettes, Polystyrene, Sterile, Plugged, Corning
Supplier: Corning
357529
53300-400CS 322.07 USD
53300-400
Falcon® Disposable Serological Pipettes, Polystyrene, Sterile, Plugged, Corning
Pipettes Serological Pipettes
Color-coded pipettes feature both negative and reverse graduations (except on 1 ml pipet).
Select pipettes are available with an extended capacity of 36 ml.
Printing is crisp, dark, and easy-to-read. Exclusive graduation alignment mark allows graduations to be brought into the line of sight quickly. Pipettes are sterile and certified nonpyrogenic.
Ordering information: Available individually wrapped in paper/plastic, individually wrapped in Advantage™ all-plastic thermoformed wrap for lower particulates, or bulk packaged.