Pure™ XLP Pipette Tips, Molecular BioProducts
Supplier: Thermo Scientific
3540-11
53225-684CS 1805.19 USD
53225-684
Pure™ XLP Pipette Tips, Molecular BioProducts
Pipette Tips
Ideal for longer tubes, these tips feature an extended length design making the bottom of 12×75 culture tubes and vials easier to reach and reducing the risk of contamination.
  • Sterile tips are certified RNase-, DNase-, DNA-, and pyrogen-free
  • Nonsterile tips are autoclavable
Tips are compatible with all major brand pipettors.
Ordering information: Tips 53225-686 and -688 are packaged 96 per tray, 8 trays per pack. Tips 53225-684 are bulk packaged.