Disposable Serological Pipettes, Glass, Sterile, Plugged, Kimble Chase
Supplier: DWK Life Sciences (KIMBLE)
72105-11100 72105-5110 72100-21100 72105-10110 72105-21100 72100-11100 72100-10110 72105-1110 72100-1110 72100-5110
14673-212CS 1032.42 USD
14673-212 14673-216 14673-224 53223-020 53223-025 14673-222 14673-228 24631-198 53223-000 53223-005
Disposable Serological Pipettes, Glass, Sterile, Plugged, Kimble Chase
Pipettes Serological Pipettes
These pipettes feature a uniform pulled tip and are clean, plugged, and ready for use.
  • Non-pyrogenic
  • Negative graduations increase versatility
Calibrated to deliver to the tip, with blowout for full rated capacity.
Certifications: Constructed from borosilicate glass in compliance with ASTM Specification E438, Type I, Class B glass requirements. Designed from ASTM Specification E714.
Ordering information: Color-coding indicates the packaging color.