Nalgene® Dropper Pipette, Low-Density Polyethylene, Thermo Scientific
Supplier: Thermo Fisher Scientific
6219-0068
52950-206CS 642.4 USD
52950-206
Nalgene® Dropper Pipette, Low-Density Polyethylene, Thermo Scientific
Pipettes Pasteur/Transfer Pipettes
Molded in one piece construction.
  • Chemically compatible with most acids, bases and alcohols
  • Nonwetting tapered tip delivers 30 to 35 drops per ml
  • Visible liquid level
The dropper bulb provides excellent drop formation. This low density polyethylene dropper is safer than glass droppers and economical.
Tube length: 68 mm