Viscosity Tubes, Cargille Laboratories
Supplier: Cargille Laboratories
44106
52944-300CS 596.77 USD
52944-300
Viscosity Tubes, Cargille Laboratories
Viscometers Capillary Viscometers
Viscosity tubes are used principally for direct comparisons to Gardner Viscosity Standards.
  • With 0.65 mm wall thickness
  • Tolerance level ID: ±0.025
  • Viscosity tube is composed of soda-lime glass with flat-blottoms
  • Score marks are measured from the center outside bottom of the tubes
Viscosity tubes are permanently marked with two score marks, at 100mm and 108mm heights. The score mark at 100 mm represents the filling level and the 108 mm score is used to fix the size of the air bubble in the tube by inserting a cork to this mark.
For bubble rate rise comparisons to Gardner Viscosity Standards, (10.65) it is equivalent to Gardner A tubes.