Penetration Needle, Koehler
Supplier: Koehler
K20500-00000
52934-662EA 379.91 USD
52934-662
Penetration Needle, Koehler
Penetrometer Needles
For testing of bituminous materials
Made from hardened stainless steel with a brass shank.
Certifications: Meets ASTM D5 specifications.