Penetration Needle, Humboldt
Supplier: Humboldt Manufacturing
H-1280
52934-120EA 682.37 USD
52934-120
Penetration Needle, Humboldt
Penetrometer Needles
For penetration test of bituminous materials as specified in ASTM Method D5
Hardened stainless steel, with brass shank. Exposed needle length: 40–45mm (11/2–13/4").