LPG Corrosion Test Cylinder, Koehler
Supplier: Koehler
K40000
52933-560EA 2745.73 USD
52933-560
LPG Corrosion Test Cylinder, Koehler
Cylinders Hydrometer Cylinders
Stainless steel cylinder with 1/4" needle valves for purging and admitting LPG samples
Conforms to ASTM D1838 and related specifications.
Dip tube with hook suspends copper strip in sample. Knurled, threaded cap with O-ring gasket hand tightens to a positive seal. Withstands hydrostatic test pressure of 1000psig (6895kPa).