Cork Borer Sharpener, Humboldt
Supplier: Humboldt Manufacturing
H-9680
52924-950EA 62.5 USD
52924-950
Cork Borer Sharpener, Humboldt
Cork Borers
The handy tool is used to sharpen borers from 3/16" to 15/16" (5 mm to 24 mm)
The solid brass cone has a hinged steel blade in a slot, and is mounted in a hardwood handle.