Saybolt Chromometer, Koehler Instrument
Supplier: Koehler
K13009
52924-046EA 10605.77 USD
52924-046
Saybolt Chromometer, Koehler Instrument
Colorimeters Comparator Kits
The Saybolt Chromometer measures color by comparing a column of sample against standard color discs.
  • Three-position color standard turret
  • Tests non-fluid waxes and liquid petroleum products
Determines Saybolt Color of highly refined petroleum products. Consists of a matched set of sample and standard tube assemblies with optical viewer. Compares a sample of the product to be tested against standard color discs under a uniform light source. Reduce column height until the sample field is lighter than the color standard and convert height to Saybolt Color using chart on instrument. Three-position turret on standard tube permits convenient changing of color disc combinations.
Certifications: Conforms to ASTM D156; DIN 51411; FTM 791-101; NF M 07-003.
Accessories information: Accessory Daylight Lamp (Model No. K13010) provides standard light source per ASTM specifications.
Ordering information: includes three whole color standards, one half color standard, and a engraved conversion chart.