PYREX® Ramsbottom Carbon Residue Bulb, Corning
Supplier: Corning
7735-24
52922-164CS 577.39 USD
52922-164
PYREX® Ramsbottom Carbon Residue Bulb, Corning
Petroleum Testing Apparatus
PYREX® Ramsbottom carbon residue bulbs with inlet and outlet tubes.
  • Narrow mouth neck, ground top style
  • Interchangeable stopper closes both inlet and outlet tubes
  • Used for the absorption of CO₂ in carbon determination
Tubulation Ext.Ø: 8 mm
Certifications: ASTM D-524