Diamond Scriber
Supplier: Musco Sports Lighting
200-22A-100
52865-005EA 80.37 USD
52865-005
Diamond Scriber
Pens Stylus and Scribers
For permanently marking glass slides, test tubes, and laboratory glassware
Sharp, selected diamond mounted in a round, clear aluminum handle tapered to fit hand comfortably. Tip retracts for protection and safety.
Ordering information: Also available in blue, black, red, and silver handles; contact your Avantor sales representative for more information.