Diversified Biotech Synergel™ Agarose Additive
Supplier: Diversified Biotech
SYN-100
52857-970EA 271.26 USD
52857-970
Diversified Biotech Synergel™ Agarose Additive
Electrophoresis Gels Agarose and Acrylamide
Better sieving than agarose, 1 gram Synergel = 2 grams agarose.High Resolution
Synergel/Agarose gels exhibit improved spatial separation and tighter banding for DNA fragments of 50-2000bp. No Background Fluorescence. The superior optical clarity permits high quality photo-documentation. Increased Load Capacity—Accommodates greater quantities of DNA reducing band tailing and smearing. Easy To Use; mix, boil, and pour.