Sight Savers® Premoistened Lens Cleaning Tissues, Bausch & Lomb®
Supplier: Bausch & Lomb
8574GM
52848-078CS 296.37 USD
52848-078
Sight Savers® Premoistened Lens Cleaning Tissues, Bausch & Lomb®
Wipes Disposable Wipes and Towels Lens Papers
These nonsilicone, antistatic tissues are individually packaged and treated with special surfactants to clean all glass and plastic lenses.
Perfect for computer screens, mirrors, safety glasses, and face shields. Unique package design allows easy dispensing of large 12.7×20.3 cm (5×8") folded, premoistened tissues. Cardboard dispenser box measures 22.9×13.7×6 cm (9×5³/₈×2³/₈").