KIMAX® Viscosity Tube, Varnish, (Bubble Tube), Kimble Chase, DWK Life Sciences
Supplier: DWK Life Sciences (KIMBLE)
46463-1075
52642-000CS 789.34 USD
52642-000
KIMAX® Viscosity Tube, Varnish, (Bubble Tube), Kimble Chase, DWK Life Sciences
Viscometers Capillary Viscometers
For rough control work where bubble comparison is made to limit varnish samples of known viscosity, reference ASTM D 1545
114 mm long with 10.75 ±0.125 mm inside diameter. With durable white ceramic enamel lines at 27, 100 and 108 mm from the outside bottom.