Autoclave Bag Red with Heat Indicator 250°F, Hardy Diagnostics

Supplier: Hardy Diagnostics
AB5819
52428-868PK 155.12 USD
52428-868
Autoclave Bag Red with Heat Indicator 250°F, Hardy Diagnostics
Bags Sterilization Pouches
Autoclave bag is made of tough, thick polypropylene, double seamed bottom, solid red bag with black biohazard warnings in English and Spanish.

  • Dimension: 8×12"
  • Thickness: 2 mil
  • Bag is rated to withstand temperatures up to 250 °F

Storage temperature: 15...30 °C. Heat indicator strip turns black when exposed to steam, OSHA approved labeling, plastic twist ties for closure furnished upon request.

Packaging: 400 per case
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR