Envirene™ Xylene Substitute, for Histology and Pathology, Hardy Diagnostics
Supplier: HARDY DIAGNOSTICS
CE128
52428-854EA 122.42 USD
52428-854
Envirene™ Xylene Substitute, for Histology and Pathology, Hardy Diagnostics
Stains Microscopy and staining reagents
An odorless non-toxic substitute for xylene, can be used for any procedure that calls for xylene. For histology, parasitology, or for slide cleaning. Has no citrus smell.