Pivoting Blade Impeller, 2.4", Caframo
Supplier: Caframo
A140
52428-772EA 156.38 USD
52428-772
Pivoting Blade Impeller, 2.4", Caframo
Stirrers Stirring Tools
The Caframo Pivoting Blade Impeller features a 2.4" diameter, and is pinned to a 5/16" by 16" shaft
The impeller will vortex 5 gallons of water at 800rpm. Designed for use with narrow neck vessels.
Composed of 316L electropolished stainless steel, it offers moderate shear with moderate vortexing characteristics.