Sweep Blades, Caframo
Supplier: Caframo
SW4
52428-758PK 136.2 USD
52428-758
Sweep Blades, Caframo
Stirrers Stirring Blades
The Caframo Sweep Blade is specifically designed for mixing viscous products
Sweep Blades are molded in Silicone SCX 0570, and are FDA compliant.
The Sweep pulls the product from the side walls of a 4 liter vessel for more effective mixing. Sweep blades are designed for use with the U044 Anchor paddle.
Ordering information: The Sweep Blade can be purchased separately, or with a 4x4" Anchor paddle.