Dowex® 1X8 (Cl) (strongly basic anion exchanger), Supelco®

Supplier: MilliporeSigma

1.05242.0250
52428-658EA 436.68 USD
52428-658
Dowex® 1X8 (Cl) (strongly basic anion exchanger), Supelco®
Dowex® 1X8 (Cl)
MDL Number: MFCD00132718
CAS Number: 12627-85-9

Order Now

Specification Test Results

Density 1.040 g/cm3 (20 °C)
pH (H2O) not applicable


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR