Ion exchange chromatography media, Ion Exchanger I
Supplier: MilliporeSigma
1.04765.0500
52428-632EA 240.75 USD
52428-632 52428-634
Ion exchange chromatography media, Ion Exchanger I
Protein Purification Protein Purification Resins
Ion Exchanger I is a strong acid cation exchangers widely used in separation of noble earths and amino acids, water treatment, analysis and food industry. The bond strength of equivalent ions increases with decreasing diameter of the hydrated ion.
Maximum working temperature: 120 °C
Exchange capacity: >1,7 mval/ml