Anti-IL-25 Rat Monoclonal Antibody (Biotin) [clone: 35B]

Supplier: Biolegend
514405
514405-BLEA 198.1 USD
514405-BL
Anti-IL-25 Rat Monoclonal Antibody (Biotin) [clone: 35B]
Antibodies
Biotin anti-mouse IL-25 (IL-17E) [35B]; Isotype: Rat IgG; Reactivity: Mouse; Apps: ELISA Detection; Size: 50 μg

Each lot of this antibody is quality control tested by ELISA assay. It is recommended that the reagent be titrated for optimal performance for each application.

Type: Primary
Antigen: IL-25
Clonality: Monoclonal
Clone: 35B
Conjugation: Biotin
Epitope:
Host: Rat
Isotype: Rat IgG
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR