Anti-IL-9 Rat Monoclonal Antibody (PE (Phycoerythrin)) [clone: RM9A4]

Supplier: Biolegend
514103 514104
514103-BLEA 205.9 USD
514103-BL 10761-686
Anti-IL-9 Rat Monoclonal Antibody (PE (Phycoerythrin)) [clone: RM9A4]
Antibodies
PE anti-mouse IL-9 [RM9A4]; Isotype: Rat IgG1, κ; Reactivity: Mouse; Apps: ICFC; Size: 100 μg

Each lot of this antibody is quality control tested by intracellular immunofluorescent staining with flow cytometric analysis. For flow cytometric staining, the suggested use of this reagent is 0.25 μg per million cells in 100 μl volume. It is recommended that the reagent be titrated for optimal performance for each application.

Type: Primary
Antigen: IL-9
Clonality: Monoclonal
Clone: RM9A4
Conjugation: PE (Phycoerythrin)
Epitope:
Host: Rat
Isotype: Rat IgG1, κ
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR