Anti-IFN-b Mouse Monoclonal Antibody [clone: IFNb/A1]

Supplier: Biolegend
514002
514002-BLEA 344.82 USD
514002-BL
Anti-IFN-b Mouse Monoclonal Antibody [clone: IFNb/A1]
Antibodies
Purified anti-human IFN-β [IFNb/A1]; Isotype: Mouse IgG1, κ; Reactivity: Human; Apps: WB, ELISA; Size: 200 μg

Each lot of this antibody is quality control tested by Western blotting. Western blotting, suggested working dilution(s): Use 5 μg antibody per 5 ml antibody dilution buffer for each mini-gel. It is recommended that the reagent be titrated for optimal performance for each application.

Type: Primary
Antigen: IFN-b
Clonality: Monoclonal
Clone: IFNb/A1
Conjugation:
Epitope:
Host: Mouse
Isotype: Mouse IgG1, κ
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR