Anti-IL-32abd Mouse Monoclonal Antibody [clone: KU32-56]

Supplier: Biolegend
513601
513601-BLEA 153.1 USD
513601-BL
Anti-IL-32abd Mouse Monoclonal Antibody [clone: KU32-56]
Antibodies
Purified anti-human IL-32αβδ [KU32-56]; Isotype: Mouse IgG1, κ; Reactivity: Human; Apps: ELISA Capture, WB, IF, IP; Size: 50 μg

Each lot of this antibody is quality control tested by ELISA assay. For use as an ELISA capture antibody, a concentration range of 4-6 μg/ml is recommended. To obtain a linear standard curve, serial dilutions of human IL-32 recombinant protein ranging from 2000 to 15 pg/ml are recommended for each ELISA plate. It is recommended that the reagent be titrated for optimal performance for each application.

Type: Primary
Antigen: IL-32abd
Clonality: Monoclonal
Clone: KU32-56
Conjugation:
Epitope:
Host: Mouse
Isotype: Mouse IgG1, κ
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR