BCR® CleaNotes® Sticky Notes, Berkshire
Supplier: Berkshire
BCN.0303.24
51280-640CS 690 USD
51280-640
BCR® CleaNotes® Sticky Notes, Berkshire
Notepads
Ideal for leaving quick messages.
  • BCR® CleaNotes® are made with cleanroom white bond paper
  • Specialized coating to reduce particles and fibers
  • Smooth surface reduces surface particle contamination
  • Latex-free
  • ISO Class 3–8
  • Cleanroom Class 1–100,000
Special adhesive backing won’t leave contaminants in the cleanroom
Packaging: 100 sheets/pad, 24 pads/case.