Nalgene® Polyolefin Plastic Paper Sheets, Thermo Scientific
Supplier: Thermo Fisher Scientific
6304-9811
51280-114CS 990.81 USD
51280-114
Nalgene® Polyolefin Plastic Paper Sheets, Thermo Scientific
Paper
Nalgene® Polyolefin Plastic Paper Sheets roll or fold without cracking and stay flexible at cold temperatures
They won't rip, stretch or shrink and can be punched for use in binders or cut into labels. This paper is resistant to many solvents including acetone, formaldehyde, formalin, xylene, acetic acid and ethanol. They accept many inks including laser, inkjet, copier, markers, ballpoint, gels and technical markers. These sheets DO NOT accept pencil.