KIMAX® Connecting Bulb, Kjeldahl, Cylindrical Type, Kimble Chase, DWK Life Sciences
Supplier: DWK Life Sciences (KIMBLE)
16040-55
51150-045CS 254.62 USD
51150-045
KIMAX® Connecting Bulb, Kjeldahl, Cylindrical Type, Kimble Chase, DWK Life Sciences
Distillation Components Distillation Receivers
Inlet and outlet tubes are curved in opposite directions within the bulb
Lower tubes are 11 to 12mm O.D. to fit special Kjeldahl stoppers.